Learn More
Agilent Technologies Headspace Crimp Caps, 20 mm, Bimetal Magnetic, PTFE/Silicone Septa, 100/Pk.
Catalog No. NC3527284
Encompass
These 20 mm crimp headspace caps are designed for reliable use in automated gas chromatography systems. The bimetal magnetic construction ensures full compatibility with robotic autosamplers, while the integrated PTFE/silicone septa provide high-performance sealing and excellent chemical resistance for headspace analysis.
  • Designed for headspace GC applications
  • Magnetic construction compatible with robotic autosamplers
  • Features 20 mm bimetal magnetic caps
  • Constructed with PTFE/silicone septa for optimal sealing
  • Maximum temperature rating of 180°C
  • Supplied in a pack of 100
Catalog No. NC3527284 Supplier Agilent Technologies Supplier No. 80100420
These 20 mm crimp headspace caps are designed for reliable use in automated gas chromatography systems. The bimetal magnetic construction ensures full compatibility with robotic autosamplers, while the integrated PTFE/silicone septa provide high-performance sealing and excellent chemical resistance for headspace analysis.
  • Designed for headspace GC applications
  • Magnetic construction compatible with robotic autosamplers
  • Features 20 mm bimetal magnetic caps
  • Constructed with PTFE/silicone septa for optimal sealing
  • Maximum temperature rating of 180°C
  • Supplied in a pack of 100