Learn More
Grainger Crimp Fitting 1-1/4 I.D. 1-1/4 F BSPP
Catalog No. 502766738
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 psi and a temperature range of -40°F to +250°F. The fitting is compatible with many different hose types and sizes. It requires a hex crimping machine for proper installation. The fitting body is made from zinc-plated steel for corrosion resistance. It meets or exceeds SAE J516 standards. This fitting provides a reliable, heavy-duty connection for high-pressure hydraulic applications.
Catalog No. 50-276-6738 Supplier Grainger Supplier No. 16P786
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 psi and a temperature range of -40°F to +250°F. The fitting is compatible with many different hose types and sizes. It requires a hex crimping machine for proper installation. The fitting body is made from zinc-plated steel for corrosion resistance. It meets or exceeds SAE J516 standards. This fitting provides a reliable, heavy-duty connection for high-pressure hydraulic applications.