Learn More
Grainger Crimp Fitting 1-1/4 I.D. M45-2 M Metric
Catalog No. 502763937
Encompass_Preferred
The Grainger website offers a wide selection of crimp hydraulic hose fittings designed to connect hydraulic hoses in fluid power systems. Available in straight, 45 degree, and 90 degree configurations, these fittings come in various thread sizes and are compatible with different hose sizes. Made from durable materials like steel, stainless steel, and brass, they feature corrosion resistance, high pressure ratings up to 5000 PSI, and a crimped connection for a reliable seal. Suitable for industrial, mobile, and marine hydraulic applications, Grainger provides competitively priced fittings from trusted brands.
Catalog No. 50-276-3937 Supplier Grainger Supplier No. 16P851
The Grainger website offers a wide selection of crimp hydraulic hose fittings designed to connect hydraulic hoses in fluid power systems. Available in straight, 45 degree, and 90 degree configurations, these fittings come in various thread sizes and are compatible with different hose sizes. Made from durable materials like steel, stainless steel, and brass, they feature corrosion resistance, high pressure ratings up to 5000 PSI, and a crimped connection for a reliable seal. Suitable for industrial, mobile, and marine hydraulic applications, Grainger provides competitively priced fittings from trusted brands.