Learn More
Grainger Crimp Fitting 1-1/4 I.D. M52-2 F Metric
Catalog No. 502764565
Encompass_Preferred
The Grainger website features crimp hydraulic hose fittings with NPT connections. These fittings are designed to connect hydraulic hoses to components with NPT threaded ports. The fittings are available in a variety of sizes from -4 to -16 to accommodate different hose diameters. They are made from steel and feature zinc electroplating for corrosion resistance. The fittings use a hex crimp design for secure hose retention. They have a pressure rating up to 5,000 psi and temperature range from -40°F to 250°F. Overall, these fittings provide a reliable threaded connection method for industrial hydraulic hose systems.
Catalog No. 50-276-4565 Supplier Grainger Supplier No. 16P864
The Grainger website features crimp hydraulic hose fittings with NPT connections. These fittings are designed to connect hydraulic hoses to components with NPT threaded ports. The fittings are available in a variety of sizes from -4 to -16 to accommodate different hose diameters. They are made from steel and feature zinc electroplating for corrosion resistance. The fittings use a hex crimp design for secure hose retention. They have a pressure rating up to 5,000 psi and temperature range from -40°F to 250°F. Overall, these fittings provide a reliable threaded connection method for industrial hydraulic hose systems.