Learn More
Grainger Crimp Fitting 1/2 I.D. 1/2 M BSPP
Catalog No. 502767476
Encompass_Preferred
The Aeroquip 16P806 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and creates a leak-proof seal when properly crimped. It can be used in hydraulic applications where tubing needs to connect to threaded ports. The compact design allows tubing runs in tight spaces.
Catalog No. 50-276-7476 Supplier Grainger Supplier No. 16P806
The Aeroquip 16P806 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and creates a leak-proof seal when properly crimped. It can be used in hydraulic applications where tubing needs to connect to threaded ports. The compact design allows tubing runs in tight spaces.