Learn More
Grainger Crimp Fitting 1/2 I.D. M22-1.5 F Metric
Catalog No. 502764564
Encompass_Preferred
The Aeroquip 16P858 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 JIC 37° female outlet. It is rated for up to 5000 psi working pressure and has a maximum operating temperature of 250°F. The fitting body is zinc plated for corrosion resistance. This hydraulic crimp fitting provides a reliable connection method for high pressure hydraulic hose in industrial applications. The crimp technology allows for quick, easy hose assembly without thread sealants or wrenches.
Catalog No. 50-276-4564 Supplier Grainger Supplier No. 16P858
The Aeroquip 16P858 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 JIC 37° female outlet. It is rated for up to 5000 psi working pressure and has a maximum operating temperature of 250°F. The fitting body is zinc plated for corrosion resistance. This hydraulic crimp fitting provides a reliable connection method for high pressure hydraulic hose in industrial applications. The crimp technology allows for quick, easy hose assembly without thread sealants or wrenches.