Learn More
Grainger Crimp Fitting 1/2 I.D. M22-1.5 M Metric
Catalog No. 502764026
Encompass_Preferred
The Grainger 1/2" I.D. M22 x 1.5 M Metric Crimp Fitting is a metric crimp fitting with a 1/2" inner diameter and M22 x 1.5 M male threads. It is made from brass and is designed for use in hydraulic and pneumatic applications. The fitting has a maximum pressure rating of 5000 psi and temperature range of -65 to +275°F. It conforms to DIN 3861 standards. The crimp fitting provides a reliable leak-proof connection.
Catalog No. 50-276-4026 Supplier Grainger Supplier No. 16P847
The Grainger 1/2" I.D. M22 x 1.5 M Metric Crimp Fitting is a metric crimp fitting with a 1/2" inner diameter and M22 x 1.5 M male threads. It is made from brass and is designed for use in hydraulic and pneumatic applications. The fitting has a maximum pressure rating of 5000 psi and temperature range of -65 to +275°F. It conforms to DIN 3861 standards. The crimp fitting provides a reliable leak-proof connection.