Learn More
Grainger Crimp Fitting 1/2 I.D. M24-1.5 M Metric
Catalog No. 502764571
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2 inch trade size and is made of wrought copper. The fitting meets SAE J2051 specifications and is UL listed. It can withstand up to 300 psi of pressure at 250 degrees Fahrenheit. The crimp fitting requires a crimping tool to secure tubing. This product provides a reliable copper to copper connection.
Catalog No. 50-276-4571 Supplier Grainger Supplier No. 16P884
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2 inch trade size and is made of wrought copper. The fitting meets SAE J2051 specifications and is UL listed. It can withstand up to 300 psi of pressure at 250 degrees Fahrenheit. The crimp fitting requires a crimping tool to secure tubing. This product provides a reliable copper to copper connection.