Learn More
Grainger Crimp Fitting 1/2 I.D. M24-1.5 F Metric
Catalog No. 502764512
Encompass_Preferred
The Grainger 1/2" I.D. M24-1.5F Metric Crimp Fitting is a push-to-connect style fitting made of brass with a nickel plated finish. It has a 1/2" inner diameter and M24 x 1.5 female metric thread. This fitting creates a leak-proof connection for use in pneumatic and hydraulic applications. It is rated for a maximum pressure of 3000 psi and temperature range of -65°F to +250°F. The package contains 10 fittings. This product can provide a reliable crimp connection.
Catalog No. 50-276-4512 Supplier Grainger Supplier No. 16P859
The Grainger 1/2" I.D. M24-1.5F Metric Crimp Fitting is a push-to-connect style fitting made of brass with a nickel plated finish. It has a 1/2" inner diameter and M24 x 1.5 female metric thread. This fitting creates a leak-proof connection for use in pneumatic and hydraulic applications. It is rated for a maximum pressure of 3000 psi and temperature range of -65°F to +250°F. The package contains 10 fittings. This product can provide a reliable crimp connection.