Learn More
Grainger Crimp Fitting 1/4 I.D. 1/2 M Flare
Catalog No. 502751661
Encompass_Preferred
The Aeroquip 14L761 is a hydraulic crimp fitting made of steel with a zinc dichromate finish. It has a 1/4" NPT male inlet and a #6 SAE female outlet. The operating pressure is up to 5000 psi. This fitting creates a reliable leak-proof seal. It is designed for use in hydraulic systems in industrial equipment, vehicles, and machinery. The zinc dichromate finish resists corrosion. This fitting allows easy installation by crimping tubing onto the fitting using the proper crimping tool, eliminating the need for welding or soldering. It is an economical and durable fitting for hydraulic applications requiring a male NPT thread connection.
Catalog No. 50-275-1661 Supplier Grainger Supplier No. 14L761
The Aeroquip 14L761 is a hydraulic crimp fitting made of steel with a zinc dichromate finish. It has a 1/4" NPT male inlet and a #6 SAE female outlet. The operating pressure is up to 5000 psi. This fitting creates a reliable leak-proof seal. It is designed for use in hydraulic systems in industrial equipment, vehicles, and machinery. The zinc dichromate finish resists corrosion. This fitting allows easy installation by crimping tubing onto the fitting using the proper crimping tool, eliminating the need for welding or soldering. It is an economical and durable fitting for hydraulic applications requiring a male NPT thread connection.