Learn More
Grainger Crimp Fitting 1/4 I.D. 1/4 F Metric
Catalog No. 502764209
Encompass_Preferred
The Aeroquip 16P889 hydraulic crimp fitting is made of steel and has a 1/4" NPT female thread. It is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. The fitting has a zinc dichromate finish for corrosion resistance. It meets or exceeds SAE J514 specifications. The 16P889 crimp fitting provides a reliable, reusable connection for hydraulic tubing in fluid power systems.
Catalog No. 50-276-4209 Supplier Grainger Supplier No. 16P889
The Aeroquip 16P889 hydraulic crimp fitting is made of steel and has a 1/4" NPT female thread. It is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. The fitting has a zinc dichromate finish for corrosion resistance. It meets or exceeds SAE J514 specifications. The 16P889 crimp fitting provides a reliable, reusable connection for hydraulic tubing in fluid power systems.