Learn More
Grainger Crimp Fitting 1/4 I.D. 1/4 M BSPP
Catalog No. 502767474
Encompass_Preferred
The Aeroquip 16P803 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. This reusable crimp fitting creates a leakproof seal. It requires a hex crimping tool to properly secure tubing. The 16P803 fitting weighs 0.1 lbs and measures 1-1/4" in length. It can withstand operating temperatures from -65 to +250 degrees Fahrenheit. This hydraulic crimp fitting is manufactured in the USA by Eaton's Aeroquip division.
Catalog No. 50-276-7474 Supplier Grainger Supplier No. 16P803
The Aeroquip 16P803 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. This reusable crimp fitting creates a leakproof seal. It requires a hex crimping tool to properly secure tubing. The 16P803 fitting weighs 0.1 lbs and measures 1-1/4" in length. It can withstand operating temperatures from -65 to +250 degrees Fahrenheit. This hydraulic crimp fitting is manufactured in the USA by Eaton's Aeroquip division.