Learn More
Grainger Crimp Fitting 1/4 I.D. 1/4 M BSPT
Catalog No. 502765908
Encompass_Preferred
The Aeroquip 16P810 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting body is zinc plated for corrosion resistance. It requires a hex crimping tool to properly secure hydraulic hose ends. This fitting provides a reliable, reusable connection method for hydraulic hoses in industrial applications.
Catalog No. 50-276-5908 Supplier Grainger Supplier No. 16P810
The Aeroquip 16P810 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting body is zinc plated for corrosion resistance. It requires a hex crimping tool to properly secure hydraulic hose ends. This fitting provides a reliable, reusable connection method for hydraulic hoses in industrial applications.