Learn More
Grainger Crimp Fitting 1/4 I.D. 3/8 F BSPP
Catalog No. 502768394
Encompass_Preferred
The Grainger 16P778 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 10mm to 19mm, providing a versatile range of metric sockets for impact wrench use. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. With deep broach depths to reach fasteners in tight spaces, these impact sockets are designed for high torque applications. The blow molded case keeps the set protected and organized. Overall, this Westward socket set offers a quality selection of 3/8-inch drive metric impact sockets for tightening and loosening hardware in heavy duty tasks.
Catalog No. 50-276-8394 Supplier Grainger Supplier No. 16P778
The Grainger 16P778 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 10mm to 19mm, providing a versatile range of metric sockets for impact wrench use. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. With deep broach depths to reach fasteners in tight spaces, these impact sockets are designed for high torque applications. The blow molded case keeps the set protected and organized. Overall, this Westward socket set offers a quality selection of 3/8-inch drive metric impact sockets for tightening and loosening hardware in heavy duty tasks.