Learn More
Grainger Crimp Fitting 1/4 I.D. M12-1.5 F Metric
Catalog No. 502766877
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The crimp fitting is compatible with many different Aeroquip hose types and sizes. Key features include corrosion resistance, vibration resistance, and a compact design. The crimp system provides a reliable, reusable fitting for hydraulic applications.
Catalog No. 50-276-6877 Supplier Grainger Supplier No. 16P816
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The crimp fitting is compatible with many different Aeroquip hose types and sizes. Key features include corrosion resistance, vibration resistance, and a compact design. The crimp system provides a reliable, reusable fitting for hydraulic applications.