Learn More
Grainger Crimp Fitting 1/4 I.D. M12-1.5 M Metric
Catalog No. 502763787
Encompass_Preferred
The Aeroquip 16P842 hydraulic crimp fitting is made of steel with a zinc dichromate finish. It has a 1/4" NPT male thread and is designed for use with 1/4" OD tubing. The working pressure is up to 5000 psi. This reusable fitting creates a leakproof seal. It is RoHS compliant.
Catalog No. 50-276-3787 Supplier Grainger Supplier No. 16P842
The Aeroquip 16P842 hydraulic crimp fitting is made of steel with a zinc dichromate finish. It has a 1/4" NPT male thread and is designed for use with 1/4" OD tubing. The working pressure is up to 5000 psi. This reusable fitting creates a leakproof seal. It is RoHS compliant.