Learn More
Grainger Crimp Fitting 1/4 I.D. M14-1.5 F Metric
Catalog No. 502765318
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for secure hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The fitting body is zinc plated steel with an O-ring seal. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. The fitting requires a hex crimping machine for proper installation. Overall, this is a heavy-duty hydraulic fitting suitable for high pressure hydraulic hose connections across a wide temperature range.
Catalog No. 50-276-5318 Supplier Grainger Supplier No. 16P817
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for secure hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The fitting body is zinc plated steel with an O-ring seal. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. The fitting requires a hex crimping machine for proper installation. Overall, this is a heavy-duty hydraulic fitting suitable for high pressure hydraulic hose connections across a wide temperature range.