Learn More
Grainger Crimp Fitting 1/4 I.D. M16-1.5 M Metric
Catalog No. 502764492
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with hoses and tubing in hydraulic applications. The crimp fitting creates a strong, reliable seal that is vibration and impulse resistant. It is available in sizes from -04 to -16 to accommodate a range of hose diameters. The fitting meets or exceeds SAE J1453 and works with many common hydraulic hoses. Overall, it provides a durable, heavy-duty crimped connection for hydraulic systems.
Catalog No. 50-276-4492 Supplier Grainger Supplier No. 16P844
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with hoses and tubing in hydraulic applications. The crimp fitting creates a strong, reliable seal that is vibration and impulse resistant. It is available in sizes from -04 to -16 to accommodate a range of hose diameters. The fitting meets or exceeds SAE J1453 and works with many common hydraulic hoses. Overall, it provides a durable, heavy-duty crimped connection for hydraulic systems.