Learn More
Grainger Crimp Fitting 1/4 I.D. M16-1.5 F Metric
Catalog No. 502766780
Encompass_Preferred
The Aeroquip 16P818 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused up to 2 times if properly crimped. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly and disassembly of hydraulic tubing systems.
Catalog No. 50-276-6780 Supplier Grainger Supplier No. 16P818
The Aeroquip 16P818 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused up to 2 times if properly crimped. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly and disassembly of hydraulic tubing systems.