Learn More
Grainger Crimp Fitting 1/4 I.D. M16-1.5 F Metric
Catalog No. 502764139
Encompass_Preferred
The Aeroquip 16P852 is a hydraulic crimp fitting made of steel with a zinc dichromate finish. It has a 1/2" NPT male thread and is designed for use with 1/2" OD tubing. The operating pressure is up to 5000 psi. The body design allows tubing to pass through the fitting and connects using a hex crimp ring and collar (sold separately). It is recommended for hydraulic applications using petroleum-based fluids. The fitting is made in the USA.
Catalog No. 50-276-4139 Supplier Grainger Supplier No. 16P852
The Aeroquip 16P852 is a hydraulic crimp fitting made of steel with a zinc dichromate finish. It has a 1/2" NPT male thread and is designed for use with 1/2" OD tubing. The operating pressure is up to 5000 psi. The body design allows tubing to pass through the fitting and connects using a hex crimp ring and collar (sold separately). It is recommended for hydraulic applications using petroleum-based fluids. The fitting is made in the USA.