Learn More
Grainger Crimp Fitting 1/4 I.D. M16-1.5 M Metric
Catalog No. 502763939
Encompass_Preferred
The Aeroquip 16P879 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. This reusable fitting creates a leakproof seal. It requires a hex crimping tool to properly secure tubing. The dimensions are 15/32" width x 1-1/32" length. This fitting is commonly used in hydraulic systems in industrial equipment, automotive applications, and more. It meets or exceeds SAE J514 specifications.
Catalog No. 50-276-3939 Supplier Grainger Supplier No. 16P879
The Aeroquip 16P879 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. This reusable fitting creates a leakproof seal. It requires a hex crimping tool to properly secure tubing. The dimensions are 15/32" width x 1-1/32" length. This fitting is commonly used in hydraulic systems in industrial equipment, automotive applications, and more. It meets or exceeds SAE J514 specifications.