Learn More
Grainger Crimp Fitting 1/4 I.D. M18-1.5 M Metric
Catalog No. 502764569
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2 inch trade size and is made of wrought copper. The fitting meets SAE J2051 specifications and is UL classified in the US and Canada. It can withstand up to 300 psi of pressure at 250 degrees Fahrenheit. The crimp fitting creates a strong, reliable, leak-free connection and is commonly used in refrigeration, air conditioning, and hydronic heating applications.
Catalog No. 50-276-4569 Supplier Grainger Supplier No. 16P880
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2 inch trade size and is made of wrought copper. The fitting meets SAE J2051 specifications and is UL classified in the US and Canada. It can withstand up to 300 psi of pressure at 250 degrees Fahrenheit. The crimp fitting creates a strong, reliable, leak-free connection and is commonly used in refrigeration, air conditioning, and hydronic heating applications.