Learn More
Grainger Crimp Fitting 1/4 I.D. M18-1.5 F Metric
Catalog No. 502764563
Encompass_Preferred
The Aeroquip 16P853 hydraulic crimp fitting is made of steel and has a 1/4" NPT female thread. It is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. The fitting has a zinc dichromate finish for corrosion resistance. It meets or exceeds SAE J514 specifications. The 16P853 crimp fitting provides a reliable, reusable connection for hydraulic applications.
Catalog No. 50-276-4563 Supplier Grainger Supplier No. 16P853
The Aeroquip 16P853 hydraulic crimp fitting is made of steel and has a 1/4" NPT female thread. It is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. The fitting has a zinc dichromate finish for corrosion resistance. It meets or exceeds SAE J514 specifications. The 16P853 crimp fitting provides a reliable, reusable connection for hydraulic applications.