Learn More
Grainger Crimp Fitting 1/4 I.D. M20-1.5 F Metric
Catalog No. 502764706
Encompass_Preferred
The Aeroquip 16P854 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be used with a wide variety of hydraulic tubing materials. This compact crimp fitting provides a reliable, heavy duty connection for hydraulic applications.
Catalog No. 50-276-4706 Supplier Grainger Supplier No. 16P854
The Aeroquip 16P854 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be used with a wide variety of hydraulic tubing materials. This compact crimp fitting provides a reliable, heavy duty connection for hydraulic applications.