Learn More
Grainger Crimp Fitting 1 I.D. 1 F BSPP
Catalog No. 502766641
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The crimp fitting body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -16 to accommodate a wide range of hose diameters. This fitting allows for a reliable, reusable connection and is commonly used in mobile and industrial hydraulic applications.
Catalog No. 50-276-6641 Supplier Grainger Supplier No. 16P785
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The crimp fitting body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -16 to accommodate a wide range of hose diameters. This fitting allows for a reliable, reusable connection and is commonly used in mobile and industrial hydraulic applications.