Learn More
Grainger Crimp Fitting 1 I.D. 1 M BSPT
Catalog No. 502766403
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is designed for use in hydraulic systems with operating pressures up to 5000 PSI and temperatures up to 250°F. The body is made from zinc plated steel for corrosion resistance. It has a hex nut for tightening and an O-ring seal. The crimp sleeve is zinc electroplated steel. This fitting connects to hydraulic hoses sized from -4 to -16. Overall, this is a high quality hydraulic crimp fitting made in the USA by Eaton that enables secure hose connections in hydraulic systems.
Catalog No. 50-276-6403 Supplier Grainger Supplier No. 16P814
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is designed for use in hydraulic systems with operating pressures up to 5000 PSI and temperatures up to 250°F. The body is made from zinc plated steel for corrosion resistance. It has a hex nut for tightening and an O-ring seal. The crimp sleeve is zinc electroplated steel. This fitting connects to hydraulic hoses sized from -4 to -16. Overall, this is a high quality hydraulic crimp fitting made in the USA by Eaton that enables secure hose connections in hydraulic systems.