Learn More
Grainger Crimp Fitting 1 I.D. M36-2 F Metric
Catalog No. 502764947
Encompass_Preferred
The Aeroquip 834GG4 is a crimp fitting made of steel with a zinc dichromate finish. It has a 1/4" tube size and is designed for use with AQP and AGS hose. The fitting has a 37° JIC male thread and is rated up to 3000 psi working pressure. This fitting allows easy assembly by crimping the ferrule onto the hose using the proper crimping tool. It provides a reliable, reusable connection for hydraulic applications. The zinc dichromate finish resists corrosion.
Catalog No. 50-276-4947 Supplier Grainger Supplier No. 16P824
The Aeroquip 834GG4 is a crimp fitting made of steel with a zinc dichromate finish. It has a 1/4" tube size and is designed for use with AQP and AGS hose. The fitting has a 37° JIC male thread and is rated up to 3000 psi working pressure. This fitting allows easy assembly by crimping the ferrule onto the hose using the proper crimping tool. It provides a reliable, reusable connection for hydraulic applications. The zinc dichromate finish resists corrosion.