Learn More
Grainger Crimp Fitting 1 I.D. M36-2 M Metric
Catalog No. 502764493
Encompass_Preferred
The Aeroquip 16P850 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #8 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi and temperatures from -40°F to +250°F. The fitting has a zinc dichromate finish for corrosion resistance. It requires a hex crimping tool to properly secure the hose. This fitting provides a reliable connection method for attaching 1/4" NPT threaded components to #8 SAE hydraulic hose.
Catalog No. 50-276-4493 Supplier Grainger Supplier No. 16P850
The Aeroquip 16P850 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #8 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi and temperatures from -40°F to +250°F. The fitting has a zinc dichromate finish for corrosion resistance. It requires a hex crimping tool to properly secure the hose. This fitting provides a reliable connection method for attaching 1/4" NPT threaded components to #8 SAE hydraulic hose.