Learn More
Grainger Crimp Fitting 1 I.D. M42-2 F Metric
Catalog No. 502763789
Encompass_Preferred
The Aeroquip 16P863 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The body is zinc plated for corrosion resistance. This reusable crimp fitting creates a reliable leak-proof seal. It requires the use of a hex crimping tool to secure special crimp sleeves over the hose fitting and hose. This fitting connects 1/4" NPT male threaded ports to #6 SAE hydraulic hose.
Catalog No. 50-276-3789 Supplier Grainger Supplier No. 16P863
The Aeroquip 16P863 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The body is zinc plated for corrosion resistance. This reusable crimp fitting creates a reliable leak-proof seal. It requires the use of a hex crimping tool to secure special crimp sleeves over the hose fitting and hose. This fitting connects 1/4" NPT male threaded ports to #6 SAE hydraulic hose.