Learn More
Grainger Crimp Fitting 3/4 I.D. 3/4 M BSPT
Catalog No. 502767096
Encompass_Preferred
The Aeroquip 16P890 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting body is zinc plated for corrosion resistance. It can be used to adapt NPT threaded ports to allow connection of #6 SAE hydraulic hose. This fitting requires proper crimping to SAE specifications for secure hose retention.
Catalog No. 50-276-7096 Supplier Grainger Supplier No. 16P813
The Aeroquip 16P890 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 SAE female outlet. It is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting body is zinc plated for corrosion resistance. It can be used to adapt NPT threaded ports to allow connection of #6 SAE hydraulic hose. This fitting requires proper crimping to SAE specifications for secure hose retention.