Learn More
Grainger Crimp Fitting 3/4 I.D. 3/4 M BSPP
Catalog No. 502764864
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" trade size and is made of wrought copper. The fitting can withstand up to 300 psi of pressure and temperatures up to 250°F. It conforms to ASME B16.22 and ASTM B75/B75M standards. The fitting has a HNBR O-ring seal and is NSF/ANSI 61 and NSF/ANSI 372 certified for potable water applications. It is manufactured in the United States.
Catalog No. 50-276-4864 Supplier Grainger Supplier No. 16P807
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" trade size and is made of wrought copper. The fitting can withstand up to 300 psi of pressure and temperatures up to 250°F. It conforms to ASME B16.22 and ASTM B75/B75M standards. The fitting has a HNBR O-ring seal and is NSF/ANSI 61 and NSF/ANSI 372 certified for potable water applications. It is manufactured in the United States.