Learn More
Grainger Crimp Fitting 3/4 I.D. M30-2 F Metric
Catalog No. 502765368
Encompass_Preferred
The Grainger 16P823 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face mounting. It can provide 0.56 kW of output power. The 16P823 is built for general purpose applications requiring up to 3/4 horsepower.
Catalog No. 50-276-5368 Supplier Grainger Supplier No. 16P823
The Grainger 16P823 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face mounting. It can provide 0.56 kW of output power. The 16P823 is built for general purpose applications requiring up to 3/4 horsepower.