Learn More
Grainger Crimp Fitting 3/4 I.D. M30-2 M Metric
Catalog No. 502764688
Encompass_Preferred
The Grainger 3/4" I.D. M30-2-M Metric Crimp Fitting is a push-to-connect style fitting made of brass with a nickel plated finish. It has a 3/4" inner diameter and is compatible with metric tubing that has a 30mm outer diameter. This fitting creates a leak-proof connection by crimping onto the tubing when inserted. It can handle pressures up to 3000 psi and temperatures from -40°F to 250°F. This fitting is commonly used in hydraulic, pneumatic, and industrial applications.
Catalog No. 50-276-4688 Supplier Grainger Supplier No. 16P886
The Grainger 3/4" I.D. M30-2-M Metric Crimp Fitting is a push-to-connect style fitting made of brass with a nickel plated finish. It has a 3/4" inner diameter and is compatible with metric tubing that has a 30mm outer diameter. This fitting creates a leak-proof connection by crimping onto the tubing when inserted. It can handle pressures up to 3000 psi and temperatures from -40°F to 250°F. This fitting is commonly used in hydraulic, pneumatic, and industrial applications.