Learn More
Grainger Crimp Fitting 3/4 I.D. M30-2 M Metric
Catalog No. 502763925
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. This fitting allows quick connect and disconnect of hydraulic hoses without thread damage. It requires a hex crimping tool to secure the hose. The compact design provides a reliable seal while minimizing space requirements.
Catalog No. 50-276-3925 Supplier Grainger Supplier No. 16P849
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. This fitting allows quick connect and disconnect of hydraulic hoses without thread damage. It requires a hex crimping tool to secure the hose. The compact design provides a reliable seal while minimizing space requirements.