Learn More
Grainger Crimp Fitting 3/8 I.D. 1/2 F BSPP
Catalog No. 502764979
Encompass_Preferred
The Aeroquip 16P780 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and features an O-ring seal. It can be used to connect hydraulic tubing in fluid power systems. The fitting is zinc plated for corrosion resistance. It meets or exceeds SAE J514 specifications.
Catalog No. 50-276-4979 Supplier Grainger Supplier No. 16P780
The Aeroquip 16P780 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and features an O-ring seal. It can be used to connect hydraulic tubing in fluid power systems. The fitting is zinc plated for corrosion resistance. It meets or exceeds SAE J514 specifications.