Learn More
Grainger Crimp Fitting 3/8 I.D. 3/8 M BSPP
Catalog No. 502766013
Encompass_Preferred
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Series 55 design. It is rated for a working pressure of 5000 PSI and a burst pressure of 25000 PSI. The fitting has a 1/4" tube OD size and is zinc yellow chromate plated. It uses a 37 degree flare type connection and is designed for use in hydraulic applications. The fitting body type is straight.
Catalog No. 50-276-6013 Supplier Grainger Supplier No. 16P804
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Series 55 design. It is rated for a working pressure of 5000 PSI and a burst pressure of 25000 PSI. The fitting has a 1/4" tube OD size and is zinc yellow chromate plated. It uses a 37 degree flare type connection and is designed for use in hydraulic applications. The fitting body type is straight.