Learn More
Grainger Crimp Fitting 3/8 I.D. 3/8 M BSPT
Catalog No. 502766766
Encompass_Preferred
The Aeroquip 16P811 hydraulic crimp fitting is a straight fitting made of steel with a zinc dichromate finish. It has a 1/4" NPT male thread and is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. This reusable fitting creates a leakproof seal. It requires a hex crimping tool to secure tubing in place. The dimensions are 15/32" width x 1-1/32" length. This fitting is commonly used in fluid power systems.
Catalog No. 50-276-6766 Supplier Grainger Supplier No. 16P811
The Aeroquip 16P811 hydraulic crimp fitting is a straight fitting made of steel with a zinc dichromate finish. It has a 1/4" NPT male thread and is designed for use with 1/4" OD tubing. The operating pressure is up to 5000 psi. This reusable fitting creates a leakproof seal. It requires a hex crimping tool to secure tubing in place. The dimensions are 15/32" width x 1-1/32" length. This fitting is commonly used in fluid power systems.