Learn More
Grainger Crimp Fitting 3/8 I.D. 5/8 M Flare
Catalog No. 502751617
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. This fitting allows quick connect and disconnect of hydraulic hoses without thread damage. It requires a hex crimping tool to securely crimp the fitting collar onto the hose. Overall, this is a heavy-duty hydraulic crimp fitting made in the USA by Eaton. It provides a reusable and durable connection for hydraulic systems.
Catalog No. 50-275-1617 Supplier Grainger Supplier No. 14L763
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body is made of carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. This fitting allows quick connect and disconnect of hydraulic hoses without thread damage. It requires a hex crimping tool to securely crimp the fitting collar onto the hose. Overall, this is a heavy-duty hydraulic crimp fitting made in the USA by Eaton. It provides a reusable and durable connection for hydraulic systems.