Learn More
Grainger Crimp Fitting 3/8 I.D. M16-1.5 F Metric
Catalog No. 502765364
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" nominal pipe size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It conforms to ASTM B75 and ASTM B88 standards. The crimp fitting is designed for use with copper tubing in potable water, oil, natural gas, steam, compressed air, and refrigerant applications. This product can be crimped using industry standard crimping tools.
Catalog No. 50-276-5364 Supplier Grainger Supplier No. 16P819
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" nominal pipe size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It conforms to ASTM B75 and ASTM B88 standards. The crimp fitting is designed for use with copper tubing in potable water, oil, natural gas, steam, compressed air, and refrigerant applications. This product can be crimped using industry standard crimping tools.