Learn More
Grainger Crimp Fitting 3/8 I.D. M16-1.5 M Metric
Catalog No. 502764025
Encompass_Preferred
The Aeroquip 16P845 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly of hydraulic lines using crimp technology.
Catalog No. 50-276-4025 Supplier Grainger Supplier No. 16P845
The Aeroquip 16P845 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly of hydraulic lines using crimp technology.