Learn More
Grainger Crimp Fitting 3/8 I.D. M18-1.5 F Metric
Catalog No. 502763938
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body material is carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. The fitting meets or exceeds SAE J516 and ISO 8434-1 standards for hydraulic hose fittings. It provides a reliable, heavy duty connection for high pressure hydraulic applications.
Catalog No. 50-276-3938 Supplier Grainger Supplier No. 16P855
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -40°F to +250°F. The body material is carbon steel with a zinc dichromate finish. It is available in sizes -04 through -32 to accommodate a wide range of hose diameters. The fitting meets or exceeds SAE J516 and ISO 8434-1 standards for hydraulic hose fittings. It provides a reliable, heavy duty connection for high pressure hydraulic applications.