Learn More
Grainger Crimp Fitting 3/8 I.D. M18-1.5 M Metric
Catalog No. 502763797
Encompass_Preferred
The Eaton Aeroquip 16P885 hydraulic crimp fitting is made of steel with a zinc dichromate finish. It has a 37° JIC 37° flare straight thread o-ring boss port, and is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting has a size of -16 (1/2" tube OD) and is compatible with tubes, hoses, and hose fittings using this size. It can be installed by crimping with the proper crimping machine and die set. This fitting provides a reliable, reusable connection method for hydraulic lines.
Catalog No. 50-276-3797 Supplier Grainger Supplier No. 16P881
The Eaton Aeroquip 16P885 hydraulic crimp fitting is made of steel with a zinc dichromate finish. It has a 37° JIC 37° flare straight thread o-ring boss port, and is designed for use in hydraulic systems with operating pressures up to 5000 psi. The fitting has a size of -16 (1/2" tube OD) and is compatible with tubes, hoses, and hose fittings using this size. It can be installed by crimping with the proper crimping machine and die set. This fitting provides a reliable, reusable connection method for hydraulic lines.