Learn More
Grainger Crimp Fitting 3/8 I.D. M20-1.5 F Metric
Catalog No. 502764686
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with 1/4" to 1" ID hoses. The fitting has a working pressure of up to 5000 psi and operating temperatures between -40°F to +250°F. It requires a hex crimping tool to properly secure the hose. This fitting allows for easy hose assembly and secure leak-proof connections. The carbon steel material provides durability while the zinc plating resists corrosion.
Catalog No. 50-276-4686 Supplier Grainger Supplier No. 16P856
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with 1/4" to 1" ID hoses. The fitting has a working pressure of up to 5000 psi and operating temperatures between -40°F to +250°F. It requires a hex crimping tool to properly secure the hose. This fitting allows for easy hose assembly and secure leak-proof connections. The carbon steel material provides durability while the zinc plating resists corrosion.