Learn More
Grainger Crimp Fitting 3/8 I.D. M20-1.5 M Metric
Catalog No. 502764570
Encompass_Preferred
The Aeroquip 16P882 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly of hydraulic lines using crimp technology. The steel construction makes it durable and suitable for industrial applications. Overall, it is a quality hydraulic crimp fitting for use with 1/4" tubing in high pressure hydraulic systems.
Catalog No. 50-276-4570 Supplier Grainger Supplier No. 16P882
The Aeroquip 16P882 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be reused. It requires a hex crimping tool to properly secure tubing. This fitting allows easy assembly of hydraulic lines using crimp technology. The steel construction makes it durable and suitable for industrial applications. Overall, it is a quality hydraulic crimp fitting for use with 1/4" tubing in high pressure hydraulic systems.