Learn More
Grainger Crimp Fitting 3/8 I.D. M22-1.5 M Metric
Catalog No. 502764712
Encompass_Preferred
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Code 55CY93. It is a hydraulic crimp fitting that creates leak-proof connections. The fitting has a working pressure of 5000 psi and a temperature range of -65 to +250°F. It uses a hex crimp design and is RoHS compliant. The fitting is designed for use in hydraulic systems with hose sizes -16 through -20. It requires a hex crimp tool to properly crimp the fitting onto the hose. The fitting dimensions are 1-5/16" diameter x 1-7/8" length.
Catalog No. 50-276-4712 Supplier Grainger Supplier No. 16P883
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Code 55CY93. It is a hydraulic crimp fitting that creates leak-proof connections. The fitting has a working pressure of 5000 psi and a temperature range of -65 to +250°F. It uses a hex crimp design and is RoHS compliant. The fitting is designed for use in hydraulic systems with hose sizes -16 through -20. It requires a hex crimp tool to properly crimp the fitting onto the hose. The fitting dimensions are 1-5/16" diameter x 1-7/8" length.