Learn More
Grainger Crimp Fitting 3/8 I.D. M22-1.5 F Metric
Catalog No. 502764027
Encompass_Preferred
The Crimp Fitting 3/8" I.D. M22-1.5 F Metric is a brass compression fitting with a 3/8" inner diameter and M22 x 1.5 female metric thread. It is made of forged brass and is nickel plated. The fitting accepts 3/8" OD tubing and is designed for use in hydraulic and pneumatic applications. It provides a leak-proof connection by compressing a sleeve against the tubing when tightened. The maximum operating pressure is 5000 psi and the maximum operating temperature is 232°C. This metric compression fitting allows easy installation by hand tightening.
Catalog No. 50-276-4027 Supplier Grainger Supplier No. 16P857
The Crimp Fitting 3/8" I.D. M22-1.5 F Metric is a brass compression fitting with a 3/8" inner diameter and M22 x 1.5 female metric thread. It is made of forged brass and is nickel plated. The fitting accepts 3/8" OD tubing and is designed for use in hydraulic and pneumatic applications. It provides a leak-proof connection by compressing a sleeve against the tubing when tightened. The maximum operating pressure is 5000 psi and the maximum operating temperature is 232°C. This metric compression fitting allows easy installation by hand tightening.