Learn More
Grainger Crimp Fitting 45 Deg 1/2 ID M22-1.5 F
Catalog No. 502766879
Encompass_Preferred
The Grainger 16P830 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.
Catalog No. 50-276-6879 Supplier Grainger Supplier No. 16P830
The Grainger 16P830 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.