Learn More
Grainger Crimp Fitting 45 Deg 1/4 ID 1/4 FBSPP
Catalog No. 502766014
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is made of steel and has a 4,000 psi working pressure. It is designed for use with AQP hose that has a 1/4" inside diameter. The fitting has a 37 degree flare seat and uses a hex crimp design. It is zinc plated for corrosion resistance. The fitting is rated for use with petroleum-based hydraulic fluids, lubrication oils, and water glycols. It meets or exceeds SAE J1453 and MIL-A-81531 standards. The fitting allows easy hose assembly and has reusability.
Catalog No. 50-276-6014 Supplier Grainger Supplier No. 16P787
The Aeroquip Hydraulic Crimp Fitting is made of steel and has a 4,000 psi working pressure. It is designed for use with AQP hose that has a 1/4" inside diameter. The fitting has a 37 degree flare seat and uses a hex crimp design. It is zinc plated for corrosion resistance. The fitting is rated for use with petroleum-based hydraulic fluids, lubrication oils, and water glycols. It meets or exceeds SAE J1453 and MIL-A-81531 standards. The fitting allows easy hose assembly and has reusability.