Learn More
Grainger Crimp Fitting 45 Deg 1/4 ID M14-1.5 F
Catalog No. 502766370
Encompass_Preferred
The Grainger 16P826 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/8" long. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It operates on 115V, 60Hz single phase power and draws 8.6 full load amps. This electric motor is suitable for a wide range of industrial, commercial and residential applications.
Catalog No. 50-276-6370 Supplier Grainger Supplier No. 16P826
The Grainger 16P826 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/8" long. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It operates on 115V, 60Hz single phase power and draws 8.6 full load amps. This electric motor is suitable for a wide range of industrial, commercial and residential applications.