Learn More
Grainger Crimp Fitting 45 Deg 1 ID M36-2 F
Catalog No. 502764485
Encompass_Preferred
The Aeroquip 16P890 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and features an O-ring seal. It can be used to make secure hydraulic connections. The fitting is zinc plated for corrosion resistance. It weighs 0.1 lbs.
Catalog No. 50-276-4485 Supplier Grainger Supplier No. 16P833
The Aeroquip 16P890 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and features an O-ring seal. It can be used to make secure hydraulic connections. The fitting is zinc plated for corrosion resistance. It weighs 0.1 lbs.